Orders LinkContactUsLinkComponent
Resonance Apparatus
Resonance Apparatus
Resonance Apparatus
Certificates
Resonance Apparatus
Resonance Apparatus
5 Options Available Below
Product Details & Documents
With the Resonance Apparatus, students investigate the effect of frequency on wavelength.
  • Durable
  • Inexpensive
  • For Determining the Wavelength and Velocity of Sound
  • Weight: 4.8 lb.
  • Height: 39" after Assembly
An 98cm, transparent, graduated plastic tube is connected to a water reservoir. Both the tube and the water reservoir are securely mounted to a metal support. The water reservoir may be raised or lowered to adjust the air column.
The apparatus weighs 4.8lbs. and is 39" when assembled. The tube is 2.5cm dia. A tuning fork is required.
Product Family Options
Información del producto
  • Availability
    Price
  • 470218-748
    Resonance Apparatus
    Resonance Apparatus
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,931.53
    $2,931.53
    Each (1UN)
       $2,931.53
    Supplier Number: PH0720
  • 470229-056
    Replacement 98 cm Graduated Tube
    Replacement 98 cm Graduated Tube
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,588.91
    $1,588.91
    Each (1UN)
       $1,588.91
    Supplier Number: PH0720TUBE
  • 470229-054
    Replacement Flexible Tubing
    Replacement Flexible Tubing
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,335.18
    $1,335.18
    Each (1UN)
       $1,335.18
    Supplier Number: PH0720FTUB
  • 470229-058
    Replacement Cup
    Replacement Cup
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $421.07
    $421.07
    Each (1UN)
       $421.07
    Supplier Number: PH0720CUP
  • 470229-052
    Resonance Apparatus, 120cm
    Resonance Apparatus, 120cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,876.56
    $3,876.56
    Each (1UN)
       $3,876.56
    Supplier Number: PH0720L