Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ wipes, flat stacked, Cellulose/polyester, 22.9×22.9 cm (9×9")
Supplier: CONTEC, INC. MX
Número de proveedor #: AMLA0001
Jump to:
Product Details & Documents
Amplitude™ Lambda™ Wipes are a hydroentangled blend of 58% cellulose and 42% polyester. The Lambda™ wipe is highly sorbent, has excellent durability, and good cleanliness.
- Hydroentangled fabric provides low levels of particles and extractables
- Excellent for wiping and absorbing spills
- Strong and durable both wet and dry
- Good cost effective, general-purpose wipe
An excellent all-purpose wipe with outstanding sorbency, it is ideal for general-purpose cleaning and spill pick-up in biotech, pharmaceutical, and medical device industries.
Recommended for ISO Class 6 – 8 environments.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Description:Amplitude™ Lambda™ wipes, flat stacked
- Material:Cellulose/polyester
- W×L:22.9×22.9 cm (9×9")