Syringe Filters with Luer Lock Inlet, Restek
Catalog # 10059-508
Supplier: RESTEK CORP
Specifications
- Ø:25 mm
- Tamaño de poro:0.45 µm
- Color:White
- Referencia:10059-508
- Membrana:PTFE
- Nº de proveedor:26145
Specifications
About this item
Syrings filters have guaranteed leak-tight connections with luer lock inlets.
- Variety of filter types, porosities, and diameters
- Color-coded for easy identification
- Rugged polypropylene housing
- Autoclavable to 121 °C for 15 minutes
- Quantity break pricing for greater savings
- Cellulose Acetate, Nylon, PES, PVDF - hydrophilic applications
- PTFE - hydrophobic applications
Syringe filters are for laboratory use only.