Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Descripción:Electrophoresis Lab System-Class Size
- No. of Lab Groups:6
- Referencia:470190-638
- Nº de proveedor:WARD470190-638
Specifications
About this item
This inexpensive set of materials outfits your whole class for gel electrophoresis at a great price.
- Precursor to working with DNA and proteins
- Material for six lab groups
- Engaging activity for students
- Estimated lab time: 45 minutes
- Inexpensive system set
These electrophoresis dyes can be used as a precursor to working with DNA and proteins. When the colorful dyes are electrophoresed, they separate by their molecular size and charge. Students compare the electrophoresed results of the individual dyes to the dye mixture to find the missing component.
Demo Size requires 5 alkaline or rechargeable 9v batteries
Class Size requires 30 alkaline or rechargeable 9v batteries