Orders LinkContactUsLinkComponent
Clean Scrub™ Mop, Contec®
Clean Scrub™ Mop, Contec®
Clean Scrub™ Mop, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Clean Scrub™ Mop, Contec®
Clean Scrub™ Mop, Contec®
2 Options Available Below
Product Details & Documents
Contec’s Clean Scrub™ single-use mop head is wrapped with a textured polyester mesh to eliminate dirt and grime from heavily contaminated surfaces. The mop head has a core of polyester fabric and foam that provides exceptional liquid absorption and resists tearing.
  • Textured polyester mesh removes heavily soiled contamination from surfaces
  • Single-use mop head eliminates cross contamination
  • Slim profile fits into small spaces
  • Compatible with a wide range of disinfectants
This pad is fastened to a semi-rigid thermoformed plate that easily snaps onto existing QuickConnect mop frames and provides steady contact pressure on mopped surfaces. The Clean Scrub™ mop will not scratch surfaces. Compatible with most disinfectants and solutions, Clean Scrub™ is ideal for many cleaning applications. Due to the thermoformed plastic backplate, Clean Scrub™ should not be autoclaved.
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • 10017-772
    Clean Scrub™ Mop Head
    Clean Scrub™ Mop Head
    10 x 33 x 3 cm (4 x 13 x 1")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (28UN)
    Case (28UN) - $12,781.61
    $12,781.61
    Case (28UN)
       $12,781.61
    Supplier Number: VKSC0003
  • 10811-978
    Clean Scrub™ Sponge
    Clean Scrub™ Sponge
    13 x 8 x 1.5 cm (5 x 3 x 0.6")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (48UN)
    Case (48UN) - $6,196.84
    $6,196.84
    Case (48UN)
       $6,196.84
    Supplier Number: VKSC0004