Sterile Flocked Swab, with Molded Plastic Applicator & Nylon Fiber Tip
Catalog # 10755-188
Supplier: COPAN DIAGNOSTICS INC
Specifications
- Descripción:Flocked Swab, with 30 mm Breakpoint from Swab Tip
- Referencia:10755-188
- Empaquetado:Peel Pouch
- Nº de proveedor:520CS01.BX
Specifications
About this item
Flocked swabs absorb and immediately release a sample into liquid media
COPAN swabs are compatible with multiple applications and platforms, such as rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, forensics, bacteriology, and virology culture.