Viscometer Holders, Cannon
Supplier: CANNON INSTRUMENT CO
Jump to:
Acerca de este artículo
All CANNON viscometer holders fit holes 51 mm (2" ) in diameter.
- Self-aligning
- Constructed of chrome-plated machined brass
- All metal holders are provided with a handle
Plastic holders are self-aligning and hold the viscometer firmly by means of a spring clip. Rubber holders are made of neoprene rubber.
1 Options Available Below
Product Details & Documents
All CANNON viscometer holders fit holes 51 mm (2" ) in diameter.
- Self-aligning
- Constructed of chrome-plated machined brass
- All metal holders are provided with a handle
Plastic holders are self-aligning and hold the viscometer firmly by means of a spring clip. Rubber holders are made of neoprene rubber.