CleanStep™ Sticky Mat, Texwipe®
Anti-microbial contamination control mat, 30 sheets/pad; 8 pads/case, 63 cm (25"), 114 cm (45"), Grey
Catalog # 89041-772
Supplier: Texwipe
: AMA254581G
Jump to:
Product Details & Documents
CleanStep™ adhesive mats is made of high grade polyethylene and it is ideal for cleanrooms and critical areas.
- Features an antimicrobial agent to prevent bacterial growth
- Numbered corner tabs to simplify inventory and replacement
- Available in 5 sizes, 3 colors, and 30 or 60 layer options
Especificaciones
- Color:Grey
- Description:Anti-microbial contamination control mat
- Length:114 cm (45")
- Packaging:30 sheets/pad; 8 pads/case
- Width:63 cm (25")