LaboPlast® and SteriPlast® Bio Samplling Spatulas, Bürkle
Catalog # 10769-684
Supplier: BURKLE INC
Specifications
- Esterilidad:Non sterile
- Color:White
- Referencia:10769-684
- Longitud de la hoja:150 mm
- Nº de proveedor:5379-0009
Specifications
About this item
These disposable spatulas are manufactured from bio-plastic 'Green PE' which is entirely made of renewable resources.
- 'Green PE' is 100% recyclable, made without fossil raw materials
- Long, ergonomic and solid handle
- Pointed blade can be easily pierced into paper and plastic bags
- Available non sterile (LaboPlast Bio) or gamma radiation sterilised (SteriPlast Bio)
- Produced in a cleanroom environment
They are ideal for the sampling of powder, granules and pastes. Insertion depth of this bio sampling spatulas is 150 mm.