LaboPlast® Bio / SteriPlast® Bio, Sampling scoops, Bürkle
Supplier: BURKLE INC
Certificates
About this item
These disposable scoops are manufactured from bio-plastic "Green PE" which is entirely made of renewable resources
- "Green PE" is 100% recyclable, made without fossil raw materials
- Available non sterile (LaboPlast Bio) or gamma radiation sterilised (SteriPlast Bio)
- Produced in a cleanroom environment
- Individually packed
They are ideal for the sampling of powder, granules and pastes.
Certifications: Complies with EU food requirements and FDA regulations.
Specifications
- Catalog No:
- 76501-374
- 76501-376
- 10769-668
- 10769-912
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- Climate Neutral Company Certificate; Material Certificate
- Climate Neutral Company Certificate; Material Certificate
- Climate Neutral Company Certificate; Material Certificate
- Climate Neutral Company Certificate; Material Certificate
Specifications
Related Information
Frequently Bought Together