SP Wilmad-LabGlass Tube for Stalagmometer, SP Industries
Catalog # 14203-776
Supplier: SP Industries
Specifications
- Capacidad:2.5 ml
- Descripción:Stalagmometer Tube
- Referencia:14203-776
- Nº de proveedor:LG-5050-102
Specifications
About this item
Specifically designed for checking the surface tension of liquids, each tube is accurately calibrated to quickly determine surface tension by the drop count method.
- Graduated above and below the bulb
This tube allow for flexibility in sample size of most liquid baths.
Delivery: All tubes are supplied with instructions for use.