Polypropylene Closures
Polypropylene closures, 20 mm, Natural
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 73660-20
Jump to:
Product Details & Documents
Polypropylene closures for 13 and 20 mm culture tubes.
- Autoclavable
- Color coded
Documents
Especificaciones
- Color:Natural
- Description:Polypropylene closures
- For Tube O.D.:20 mm