Universal Round Lighter
Universal round lighter
Catalog # 470205-738
Supplier: FULLER G C MFG CO SE
Número de proveedor #: 3001
Jump to:
Product Details & Documents
The universal round lighter is capable of lighting any flammable gas. The lighter features a very long lifespan and provides the lowest cost per ignition of any lighter anywhere.
Especificaciones
- Description:Universal round lighter