Orders LinkContactUsLinkComponent
Solid Brass Mass Set
Solid Brass Mass Set
  470206-078
Solid Brass Mass Set
  470206-078
Solid Brass Mass Set
Solid Brass Mass Set
Solid Brass Mass Set
$51.70
  31/12/2026
Each (1UN)
Solid Brass. Accurate machined masses placed on a plastic and metal rack with handle.
Set of 12; 1 x 1g, 2 x 2g, 1 x 5g, 1 x 10g, 2 x 20g, 1 x 50g, 1 x 100g, 2 x 200g, 1 x 500g. Total 1110g.
  • Description:
    Solid Brass Mass Set