Orders LinkContactUsLinkComponent
Black Rubber Bib Apron
Black Rubber Bib Apron
Black Rubber Bib Apron
Certificates
Black Rubber Bib Apron
3 Options Available Below
Product Details & Documents
Supply Your Lab with Personal Protective Equipment
  • Basic black rubber apron
  • Perfect for labs, kitchens, or shops
  • Contains latex
Suitable for general lab work. Comfortable and flexible Black Rubber Bib Aprons are waterproof and resistant to most chemicals used by students in the lab.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 470228-490
  • 470148-674
  • 470149-220
  • Garment application:
  • Food|Laboratory
  • Food|Laboratory
  • Food|Laboratory
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • Rubber
  • Rubber
  • Rubber
Especificaciones
Product Family Options
Información del producto
  • Color
    Clothing Size
    L×W
    Availability
    Price
  • Black
    XS
    22×15"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $215.34
    $215.34
    Each (1UN)
       $215.34
    Supplier Number: RBAPXS
    Especificaciones:
    More
    Specifications
    Cat. No.
    470228-490
    Garment application
    Food|Laboratory
    Gender
    Unisex
    Fabric
    Rubber
  • Black
    S
    30×24"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $109.11
    $109.11
    Log in Para disponibilidad
    Case de 25 (1UN)
    Case de 25 (1UN) - $118.93
    $118.93
    Each (1UN)
       $109.11
    Case de 25 (1UN)
       $118.93
    Supplier Number: RBAP-CH
    Especificaciones:
    More
    Specifications
    Cat. No.
    470148-674
    Garment application
    Food|Laboratory
    Gender
    Unisex
    Fabric
    Rubber
  • Black
    M
    36×27"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $142.96
    $142.96
    Log in Para disponibilidad
    Case de 25 (1UN)
    Case de 25 (1UN) - $155.83
    $155.83
    Each (1UN)
       $142.96
    Case de 25 (1UN)
       $155.83
    Supplier Number: RBAP-MD
    Especificaciones:
    More
    Specifications
    Cat. No.
    470149-220
    Garment application
    Food|Laboratory
    Gender
    Unisex
    Fabric
    Rubber