Orders LinkContactUsLinkComponent
Polymethylpentene Cylinders
Polymethylpentene Cylinders
Supplier:   DYNALON
Polymethylpentene Cylinders
Supplier:  DYNALON
Certificates
Polymethylpentene Cylinders
Polymethylpentene Cylinders
3 Options Available Below
Product Details & Documents
Glass-Like Clarity.
  • Permanently molded graduations
  • Autoclavable
The cylinders are chemically resistant to most acids, bases, alcohols, and aldehydes.
Product Family Options
Product Information
  • Capacity
    Height
    Ø
    Graduation Range
    Tolerance
    Availability
    Price
  • 100 ml
    250 mm
    31 mm
    1 ml
    1 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $202.61
    $202.61
    Log in for availability
    Case de 10 (1UN)
    Case de 10 (1UN) - $220.84
    $220.84
    Each (1UN)
       $202.61
    Case de 10 (1UN)
       $220.84
    Supplier Number: 237765
  • 250 ml
    315 mm
    42 mm
    2 ml
    2 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $307.40
    $307.40
    Log in for availability
    Case de 8 (1UN)
    Case de 8 (1UN) - $335.07
    $335.07
    Each (1UN)
       $307.40
    Case de 8 (1UN)
       $335.07
    Supplier Number: 237775
  • 1000 ml
    440 mm
    66 mm
    10 ml
    10 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $759.95
    $759.95
    Log in for availability
    Case de 4 (1UN)
    Case de 4 (1UN) - $828.34
    $828.34
    Each (1UN)
       $759.95
    Case de 4 (1UN)
       $828.34
    Supplier Number: 237895