RIVAL™ 200 Pack Accelerator Free, Soft Modulus, Exam Grade Nitrile Gloves, TechNiGlove
About this item
Twice as many gloves in each case.
- Accelerator-free
- Powder-free
- Soft modulus
- Low extractable
- 100% nitrile
Rival's new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort. Microtextured fingertips provide excellent grip on all surfaces. Rival is 100% nitrile and contains no natural rubber. AQL level of 1.5 for pinhole protection.
Environmentally and economically conscious. All in a box saves space and reduce waste.
Ordering information: Packaged 200 gloves per box, 10 boxes per case.
3 Options Available Below
Product Details & Documents
Twice as many gloves in each case.
- Accelerator-free
- Powder-free
- Soft modulus
- Low extractable
- 100% nitrile
Rival's new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort. Microtextured fingertips provide excellent grip on all surfaces. Rival is 100% nitrile and contains no natural rubber. AQL level of 1.5 for pinhole protection.
Environmentally and economically conscious. All in a box saves space and reduce waste.
Ordering information: Packaged 200 gloves per box, 10 boxes per case.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 10791-604
- 10791-606
- 10791-608
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Rolled
- Rolled
- Rolled
- Cuff thickness:
- 3 mil
- 3 mil
- 3 mil
- Finger thickness:
- 4 mil
- 4 mil
- 4 mil
- Palm thickness:
- 3.5 mil
- 3.5 mil
- 3.5 mil
- FDA regulated:
- +
- +
- +
Specifications
Product Family Options
Product Information
- Glove sizeGlove ColorLengthThicknessAvailabilityPrice
- SBlue24.1 cm (9½")4 mil.Paquete de 200$421.35Case de 10 (200UN)$2,787.95Supplier Number: RV401Specifications:
More
Specifications
Cat. No.10791-604Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCuff styleRolledCuff thickness3 milFinger thickness4 milPalm thickness3.5 milFDA regulated+ - MBlue24.1 cm (9½")4 mil.Paquete de 200$527.67Case de 10 (200UN)$575.16Supplier Number: RV402Specifications:
More
Specifications
Cat. No.10791-606Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCuff styleRolledCuff thickness3 milFinger thickness4 milPalm thickness3.5 milFDA regulated+ - LBlue24.1 cm (9½")4 mil.Paquete de 200$491.70Case de 10 (200UN)$535.95Supplier Number: RV403Specifications:
More
Specifications
Cat. No.10791-608Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCuff styleRolledCuff thickness3 milFinger thickness4 milPalm thickness3.5 milFDA regulated+