Orders LinkContactUsLinkComponent
Plain Form API ASTM Hydrometers, Kimble Chase
Plain Form API ASTM Hydrometers, Kimble Chase
Plain Form API ASTM Hydrometers, Kimble Chase
Certificates
Plain Form API ASTM Hydrometers, Kimble Chase
1 Options Available Below
Product Details & Documents
Serial number is printed on the scale and a certificate of traceability is included
The hydrometer meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy. The certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards. The ballast is made from steel pellets and a binder, and it is free of heavier metals.
Product Family Options
Información del producto
  • Sample Volume
    Length
    Body O.D.
    Stem O.D.
    Graduation Interval
    Graduation Range
    Availability
    Price
  • 600 ml
    335 mm
    25 mm
    7 mm
    0.10° API
    API 69°-81°, ASTM 8H
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,393.03
    $1,393.03
    Each (1UN)
       $1,393.03
    Supplier Number: 52130-6981
  • 600 ml
    330 mm
    25 mm
    7 mm
    0.10° API
    API 79°-91°, ASTM 9H
    Discontinued
    Supplier Number: 52130-7991
  • 600 ml
    330 mm
    21 mm
    5 mm
    0.10° API
    API 89°-101°, ASTM 10H
    Discontinued
    Supplier Number: 52130-8910