Orders LinkContactUsLinkComponent
Precision Specific Gravity Hydrometers, Kimble Chase
Precision Specific Gravity Hydrometers, Kimble Chase
Precision Specific Gravity Hydrometers, Kimble Chase
Certificates
Precision Specific Gravity Hydrometers, Kimble Chase
Precision Specific Gravity Hydrometers, Kimble Chase
8 Options Available Below
Product Details & Documents
Serial number is printed on the scale and a certificate of traceability is included
Model 52110-2000 is a set comprising one each of all 8 precision specific gravity hydrometers.
The hydrometer meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy. The certificate of traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards. The ballast is made from steel pellets and a binder, and it is free of heavier metals.
Documents
Product Family Options
Información del producto
  • Sample Volume
    Length
    Body O.D.
    Stem O.D.
    Graduation Interval
    Graduation Range
    Availability
    Price
  • 10169-780
    Hydrometer
    Hydrometer
    300 ml
    310 mm
    16 mm
    7 mm
    0.001 SG
    SG 0.700–0.810
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
    Supplier Number: 52110-0708
  • Hydrometer
    500 ml
    300 mm
    18 mm
    7 mm
    0.001 SG
    SG 0.800–0.910
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
    Supplier Number: 52110-0809
  • 10169-784
    Hydrometer
    Hydrometer
    500 ml
    310 mm
    18 mm
    7 mm
    0.001 SG
    SG 0.900–1.010
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
    Supplier Number: 52110-0910
  • 10169-786
    Hydrometer
    Hydrometer
    500 ml
    300 mm
    16 mm
    8 mm
    0.002 SG
    SG 1.000–1.220
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
    Supplier Number: 52110-1012
  • 10169-788
    Hydrometer
    Hydrometer
    300 ml
    300 mm
    16 mm
    7 mm
    0.002 SG
    SG 1.200–1.420
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
    Supplier Number: 52110-1214
  • 10169-790
    Hydrometer
    Hydrometer
    500 ml
    300 mm
    18 mm
    7 mm
    0.002 SG
    SG 1.400–1.620
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,216.91
    $1,216.91
    Each (1UN)
       $1,216.91
    Supplier Number: 52110-1416
  • Hydrometer
    500 ml
    300 mm
    18 mm
    7 mm
    0.002 SG
    SG 1.600–1.820
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,258.63
    $1,258.63
    Each (1UN)
       $1,258.63
    Supplier Number: 52110-1618
  • 10169-796
    Hydrometer Set
    Hydrometer Set
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $18,691.06
    $18,691.06
    Each (1UN)
       $18,691.06
    Supplier Number: 52110-2000