Plate Count Agar, MilliporeSigma
EM1.05463.5000
:
- Tipo de env.:Plastic drum
- Descripción:Plate Count Agar
- Tamaño:5 kg
- Solubilidad:22.5 g/L (100 °C)
- Densidad:560 kg/m³
- Incubación:72 hrs.; 30°C; aerobic
- Almacenamiento:Store at +15°C to +25°C
- pH:7.0 (22 g/L, H₂O, 30 °C) (after autoclaving)
- Referencia:EM1.05463.5000
- Nº de proveedor:1.05463.5000
Casein-peptone glucose yeast extract agar for microbiology. Peptone from casein 5.0; Yeast extract 2.5; D(+)Glucose 1.0; Agar-agar 14.0.
- 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
- 10 to 100 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
- 10 to 100 inoculum on reference medium Escherichia coli ATCC 25922 (WDCM 00013)
- 10 to 100 inoculum on reference medium Bacillus subtilis ATCC 6633 (WDCM 00003)
- 10 to 100 inoculum on reference medium Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)
- 10 to 100 inoculum on reference medium Listeria monocytogenes ATCC 19118
- 10 to 100 inoculum on reference medium Lactobacillus acidophilus ATCC 4356 (WDCM 00098)
- 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
- 10 to 100 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
- 10 to 100 inoculum on reference medium Escherichia coli ATCC 25922 (WDCM 00013)
- 10 to 100 inoculum on reference medium Bacillus subtilis ATCC 6633 (WDCM 00003)
- Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 8739 (WDCM 00012)
- Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
- Greater than or equal to 70% recovery on test medium Bacillus subtilis ATCC 6633 (WDCM 00003)
- Greater than or equal to 70% recovery on test medium Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)
- Greater than or equal to 70% recovery on test medium Lactococcus lactis spp. lactis ATCC 19435 (WDCM 00016)
- Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 19118
- Greater than or equal to 70% recovery on test medium Lactobacillus acidophilus ATCC 4356 (WDCM 00098)
- Greater than or equal to 70% recovery on test medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
- Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 8739 (WDCM 00012)
- Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
- Greater than or equal to 70% recovery on test medium Bacillus subtilis ATCC 6633 (WDCM 00003)