MUP Selective Supplement, MilliporeSigma
MUP Selective Supplement, Fiber case, 10 x 1 vial
Supplier: MERCK, S.A. DE C.V. SV
: 1.00045.0010
MUP Selective Supplement, MilliporeSigma
Catalog # EM1.00045.0010
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
In North America 1.00045.0010 replaces and is identical to 1.00045.0017
- Less than or equal to 70% recovery on bifidobacterium animalis subsp. lactis ATCC 27536
- Less than or equal to 70% recovery on bifidobacterium breve ATCC 15700
- Less than or equal to 70% recovery on bifidobacterium longum ATCC 15707
- No growth in Lactobacillus casei ATCC 393 (WDCM 00100)
- No growth in Lactobacillus delbrueckii subsp. bulgaricus ATCC 11842 (WDCM 00102)
- No growth in Streptococcus salivarius subsp. thermophilus DSM 20259
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Fiber case
- Description:MUP Selective Supplement
- Size:10 x 1 vial
- Cat. No.:EM1.00045.0010
- Storage:Store at +2°C to +8°C
- Incubation:72 hrs. at 37°C, anaerobic