Orders LinkContactUsLinkComponent
Standard Grade Qualitative Filter Papers
Standard Grade Qualitative Filter Papers
Supplier:   VWR International
Standard Grade Qualitative Filter Papers
Supplier:  VWR International
Certificates
Standard Grade Qualitative Filter Papers
Standard Grade Qualitative Filter Papers
18 Options Available Below
Product Details & Documents
Qualitative Filter Paper is available in three standard grades.
  • Applicable to Most Science Education Applications
  • Grades One, Two, and Three Available
  • Grade One Works with Routine Applications
  • Grade Two Filters Large Samples
  • Grade Three Works with Buchner Funnels
Grade One filter papers are mainly used for routine applications such as separation of precipitates and filtering liquids. They have medium retention and flow rates. Grade Two filter papers are more retentive and absorbent than Grade One papers. Usually, Grade Two filter papers have an 8 µm particle size range and are used for filtering larger sample types such as soil.
Grade Three filter papers are approximately double the thickness of Grade One filter paper, which makes them more durable and suitable for use with Buchner funnels.
Packaging: One pack has 100 papers, and one case has 4800 papers.
Product Family Options
Información del producto
  • Grade
    Size
    Availability
    Price
  • 470162-996
    Filter Paper
    Filter Paper
    1
    90 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $56.99
    $56.99
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $62.11
    $62.11
    Paquete de 100
       $56.99
    Case de 48 (100UN)
       $62.11
  • 470163-000
    Filter paper
    Filter paper
    1
    110 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $70.96
    $70.96
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $77.35
    $77.35
    Paquete de 100
       $70.96
    Case de 48 (100UN)
       $77.35
  • 470163-002
    Filter paper
    Filter paper
    1
    125 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $82.23
    $82.23
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $89.63
    $89.63
    Paquete de 100
       $82.23
    Case de 48 (100UN)
       $89.63
  • 470163-024
    Filter paper
    Filter paper
    1
    150 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $118.79
    $118.79
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $129.48
    $129.48
    Paquete de 100
       $118.79
    Case de 48 (100UN)
       $129.48
  • 470162-956
    Filter paper
    Filter paper
    1
    185 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $182.75
    $182.75
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $199.20
    $199.20
    Paquete de 100
       $182.75
    Case de 48 (100UN)
       $199.20
  • 470162-958
    Filter paper
    Filter paper
    1
    215 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $246.17
    $246.17
    Log in Para disponibilidad
    Case de 24 (100UN)
    Case de 24 (100UN) - $268.32
    $268.32
    Paquete de 100
       $246.17
    Case de 24 (100UN)
       $268.32
  • 470162-960
    Filter paper
    Filter paper
    1
    240 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $290.20
    $290.20
    Log in Para disponibilidad
    Case de 24 (100UN)
    Case de 24 (100UN) - $316.32
    $316.32
    Paquete de 100
       $290.20
    Case de 24 (100UN)
       $316.32
  • 470162-962
    Filter paper
    Filter paper
    2
    90 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $67.19
    $67.19
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $73.23
    $73.23
    Paquete de 100
       $67.19
    Case de 48 (100UN)
       $73.23
  • 470162-964
    Filter paper
    Filter paper
    2
    110 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $82.77
    $82.77
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $90.22
    $90.22
    Paquete de 100
       $82.77
    Case de 48 (100UN)
       $90.22
  • 470162-966
    Filter paper
    Filter paper
    2
    125 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $103.72
    $103.72
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $113.06
    $113.06
    Paquete de 100
       $103.72
    Case de 48 (100UN)
       $113.06
  • 470162-968
    Filter paper
    Filter paper
    2
    150 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $138.12
    $138.12
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $150.56
    $150.56
    Paquete de 100
       $138.12
    Case de 48 (100UN)
       $150.56
  • 470163-004
    Filter paper
    Filter paper
    2
    185 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $221.97
    $221.97
    Log in Para disponibilidad
    Case de 48 (100UN)
    Case de 48 (100UN) - $241.94
    $241.94
    Paquete de 100
       $221.97
    Case de 48 (100UN)
       $241.94
  • 470163-008
    Filter paper
    Filter paper
    2
    240 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $326.24
    $326.24
    Liquidación
    Log in Para disponibilidad
    Case de 24 (100UN)
    Case de 24 (100UN) - $355.60
    $355.60
    Liquidación
    Paquete de 100
       $326.24
    Case de 24 (100UN)
       $355.60
  • 470163-010
    Filter paper
    Filter paper
    3
    90 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $116.09
    $116.09
    Log in Para disponibilidad
    Case de 24 (100UN)
    Case de 24 (100UN) - $126.53
    $126.53
    Paquete de 100
       $116.09
    Case de 24 (100UN)
       $126.53
  • 470163-012
    Filter paper
    Filter paper
    3
    110 mm
    Discontinued
  • 470163-014
    Filter paper
    Filter paper
    3
    125 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $177.89
    $177.89
    Log in Para disponibilidad
    Case de 24 (100UN)
    Case de 24 (100UN) - $193.90
    $193.90
    Paquete de 100
       $177.89
    Case de 24 (100UN)
       $193.90
  • 470163-016
    Filter paper
    Filter paper
    3
    150 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $229.49
    $229.49
    Log in Para disponibilidad
    Case de 24 (100UN)
    Case de 24 (100UN) - $250.14
    $250.14
    Paquete de 100
       $229.49
    Case de 24 (100UN)
       $250.14
  • 470163-020
    Filter paper
    Filter paper
    3
    215 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $470.27
    $470.27
    Log in Para disponibilidad
    Case de 12 (100UN)
    Case de 12 (100UN) - $512.59
    $512.59
    Paquete de 100
       $470.27
    Case de 12 (100UN)
       $512.59
  • 470163-022
    Filter paper
    Filter paper
    3
    240 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $539.07
    $539.07
    Log in Para disponibilidad
    Case de 12 (100UN)
    Case de 12 (100UN) - $587.59
    $587.59
    Paquete de 100
       $539.07
    Case de 12 (100UN)
       $587.59