Premium Square Bosshead
Premium Square Bosshead
Catalog # 470165-430
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PRBH05
Jump to:
Product Details & Documents
All Metal Construction to Carry Heavy Loads and Prevent Torque Damage
- Threaded metal one-piece clamping screws
- Able to hold flat and round objects up to 15 mm thick
- Holds rods orthogonal (90°) to one another
Especificaciones
- Description:Premium Square Bosshead