Premium Three-Finger Long Clamp, Vinyl Coated
470165-382
- Descripción:Premium Three-Finger Long Clamp, Vinyl Coated
- Referencia:470165-382
- Nº de proveedor:PRCMP07
Vinyl Lined Extension Lab Clamp
- Maximum clamp opening is 8.5 cm
- 14 cm connecting rod
Clamp constructed of powder coated steel, three vinyl coated fingers attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.