Premium Three-Prong Clamp with Dual Screw Adjustment, Vinyl Coated
Heavy-Duty, Spring Loaded Clamp Jaw.
- Made of steel and powder-coated alloy
- All metal adjustment screws
- Three vinyl coated clamp fingers
This three-prong clamp has a 6 cm maximum clamp opening and a 12.5 cm steel connecting rod.
Frequently Bought Together