Premium Two-Prong Clamp with Dual Screw Adjustment, Vinyl Coated
This durable clamp is made of powder-coated steel.
- Maximum Clamp Opening: 7 cm
- Connection Rod: 12.5 cm
- Spring-Loaded Clamp Jaw
- Two Vinyl-Coated Prongs
- Powder-Coated Steel Construction
The clamp has thread-fitted actuating joints and two vinyl-coated prongs.
A spring-loaded clamp jaw with a dual-screw adjustment can fine-tune any fitting.
Frequently Bought Together