Orders LinkContactUsLinkComponent
Heat Transfer Liquids for Circulators, Lauda
Heat Transfer Liquids for Circulators, Lauda
Heat Transfer Liquids for Circulators, Lauda
Heat Transfer Liquids for Circulators, Lauda
Heat Transfer Liquids for Circulators, Lauda
Heat transfer liquid is of crucial importance for the safe and reliable operation of thermostats.
  • These liqiuds are durable
  • Optimize thermostat life
  • Available for open and closed thermostats
Simple and safe handling, suitable for long term operation which allows highly accurate thermostating, even at extreme temperatures.
Product Family Options
For Open Thermostats
  • Description
    Volume
    Temperature Range
    Color
  • Kryo 60, silicone oil
    5 L
    –60...+60 °C (–76...+140 °F)
    Colourless
    Each (5l)
    Each (5l) - $38,527.34
    $38,527.34
    Each (5l)
       $38,527.34
     : LZB 102
  • Therm 250, silicone oil
    5 L
    50...250 °C (122...482 °F)
    Colourless
     : -