Premium Four-Prong Clamp, Cork Lined
Supplier: EISCO SCIENTIFIC LLC
Jump to:
Acerca de este artículo
Cork Lined Extension Lab Clamp
- Maximum clamp opening is 10.5 cm
- 14 cm stainless steel connecting rod
Clamp constructed of powder coated steel, four cork lined prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.
2 Options Available Below
Product Details & Documents
Cork Lined Extension Lab Clamp
- Maximum clamp opening is 10.5 cm
- 14 cm stainless steel connecting rod
Clamp constructed of powder coated steel, four cork lined prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.
Documents
Product Family Options
Información del producto
- AvailabilityPrice
- 470165-398Premium Four-Prong Clamp, Cork LinedSupplier Number: PRCMP02
- 470165-400Premium Four-Prong Clamp with Bosshead, Cork LinedSupplier Number: PRCMP02BH