Orders LinkContactUsLinkComponent
A-Shaped Base Retort Stand
A-Shaped Base Retort Stand
A-Shaped Base Retort Stand
A-Shaped Base Retort Stand
This cast iron, powder-coated, A-shaped base includes a stainless steel rod.
  • Powder-Coated
  • Heavy-Duty
  • A-Shaped
  • Multiple Sizes Available
  • Stainless Steel Rod Included
These bases can be nested in one another.
Multiple sizes are available.
Product Family Options
  • Description
    Rod Diameter
    Rod Length
    Thread Size
  • 7" Base with Stainless Steel Rod
    11 mm
    500 mm
    10 x 1.5 mm
    Each (1UN)
    Each (1UN) - $916.78
    $916.78
    Each (1UN)
       $916.78
     : STDRDAB01
  • 9" Base with Stainless Steel Rod
    11 mm
    600 mm
    10 x 1.5 mm
    Each (1UN)
    Each (1UN) - $1,127.32
    $1,127.32
    Each (1UN)
       $1,127.32
     : STDRDAB02
  • 11" Base with Stainless Steel Rod
    11 mm
    750 mm
    10 x 1.5 mm
    Each (1UN)
    Each (1UN) - $1,338.91
    $1,338.91
    Each (1UN)
       $1,338.91
     : STDRDAB03