DuPont™ Tyvek® IsoClean® Boot Covers with Gripper™ Sole
Proveedor: DUPONT SPECIALTY PRODUCTS USA, MX
Certificados
Acerca de este artículo
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Gripper™ soles provide enhanced skid-resistance and durability
- Elastic at ankles allows for a secure fit
- Elastic opening
- Coverage up to calf
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certificaciones: Sterile garments are gamma sterilized to SAL of 10-6.
Embalaje: Bulk packaged option packed in double transparent poly liners.
Frequently Bought Together