Orders LinkContactUsLinkComponent
VWR® Compact Micro Centrifuges
VWR® Compact Micro Centrifuges
VWR® Compact Micro Centrifuges
Supplier:  
Certificates
VWR® Compact Micro Centrifuges
0 Options Available Below
Product Details & Documents
These compact microcentrifuges are ideal for DNA, RNA, and temperature-sensitive samples.
  • Refrigerated Model with Pre-Cool Function Available
  • Robust and Steady Temperature Control
  • High Performance; Ideal for Sample Preparation
  • Short Run-up and Run-down Times
  • Pulse Button for Short Centrifugation Runs
Safety locking lids are equipped for biocontainment.
They are autoclavable and phenol-resistant. Operation noise made by these units is less than 58 dB, which is softer than a normal conversation.
Ordering information: Units come with a five-year manufacturer's warranty that extends the standard manufacturer's two-year warranty for an additional three years. The three-year extension is a full powertrain manufacturer's warranty on the centrifuge only. A comprehensive range of rotor options is available separately.
Product Family Options
Complementary Products
Información del producto
  • Catalog #
    Volume
    Color
    Style
    Sterility
    Maximum RCF
    Cap Color
    Cap Type
    Graduations
    Availability
    Price
  • 87003-294
    1.7 ml
    Natural
    Conical-Bottom
    Non sterile
    25000×g
    Log in Para disponibilidad
    Paquete de 500 - $349.87
  • 20170-038
    1.5 ml
    Natural
    Conical-Bottom
    Non sterile
    40000×g
    Log in Para disponibilidad
    Paquete de 500 - $233.26
  • 89000-028
    1.5 ml
    Natural
    Conical-Bottom
    Non sterile
    20000×g
    Log in Para disponibilidad
    Paquete de 500 - $211.77
  • 89004-302
    Green
    2 ml
    Clear
    Freestanding
    Screw cap
    Sterile
    Yes
    20000×g
    Log in Para disponibilidad
    Paquete de 500 - $863.69