Ward's® Slime Making Lab Activity
A Fun and Interesting Introduction to Polymers
- Interactive
- Hands-On
Toy stories and movies call it "slime" or "ooze", but this goopy substance is actually a great introduction to polymers for your science class! Students will delve into an overview over the kinds, uses, and examples of polymers, their disposal, and biodegradability. Then your students get real "hands-in" experience of just what polymers are, using two non-toxic chemicals to make slime.
: Includes Guar Gum, Borax Solution, Food Coloring, Pipettes, Beakers, and Wooden Stirrers.
- Description:Ward's® Slime Making Activity
- No. of Lab Groups:15
- Includes:Teacher's Guide and Student Copymaster
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.