GHS Pictogram Wallchart
GHS Pictogram Wallchart, 20×29"
Catalog # 470165-236
Supplier: EMEDCO
Número de proveedor #: WC-GHS
Jump to:
Product Details & Documents
This eye-catching wallchart illustrates GHS pictograms and their corresponding hazard classes.
- Durable 0.02" thick plastic construction
- Easy-to-read
- Banana plug clip wires provided
- Message reminds employees of potential risks
The wallchart is perfect for display in a laboratory prep room or other busy areas, so all employees are reminded of potential risks and how to identify them.
Provided banana plug clip wires can attach the poster to posts.
Especificaciones
- Description:GHS Pictogram Wallchart
- Dimensions:20×29"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



