Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Descripción:Ink Chromatography and Forensics STEM Kit
- No. of Students:40
- Referencia:470213-358
- Nº de proveedor:AISCRKIT
Students solve the case with ink chromatography and STEM.
- Hands-On STEM Activity
- Model and Inquiry Activities
- Teacher and Student Guides Included
- Material Accommodates up to 40 Students
- Covered Concepts Include Data Analysis, Chromatographs, and More
This kit introduces students to ink chromatography and its application in solving forensics cases. They gain experience in creating ink databases, identifying ‘ink tags’ to ‘date’ various inks and then use them, as professional document examiners do, in solving two mysteries involving document inks—an apparent suicide note and a forgery case.
Covered concepts include data analysis, chromatographs, inks and their composition-mixtures, compounds, mixtures, molecules, solubility, affinity, polar substances, Rf values, capillary action, light energy, fluorescence, and hypothesis testing.