Miniature LED Lights
Minature LED bulb, pack of 10, E10 base, 3.2 V
Catalog # 470224-034
Supplier: SKYWILL FAR EAST INDUSTRIES
Número de proveedor #: OPDT-W4DK86A1B
Jump to:
Product Details & Documents
These lights are great to use with a variety of electricity and science projects.
Your batteries will last longer: eco-friendly LEDs draw only 25 mA
- Bulbs light only when current is in correct direction
- Size E10 screw base
- Pack of 10
Perfect for student investigations of DC circuits. Not recommended to exceed 3 V. The bulbs light only when the current is in the correct direction.
Packaging: One package has 10 bulbs.
Especificaciones
- Description:Minature LED bulb, pack of 10, E10 base, 3.2 V