Seismic Accelerator
Seismic Accelerator
470163-912
: 7050-00
Check out this cool product!
- Safety goggles and extra balls included
- Contained in retail packaging
- Comes with a Teacher's guide
Watch the red ball rockets into the air up to five times the original drop height! Explore the Law of the Conservation of Energy with this educational but fun product!
- Description:Seismic Accelerator