Radiation Cans
Radiation Cans, Set of 3
Catalog # 470163-902
Supplier: AMERICAN SCIENTIFIC LLC SE
Número de proveedor #: AR-725
Jump to:
Product Details & Documents
Radiation cans for demonstrating solar energy and heat transfer.
- The cans are finished with silver, black, and white exteriors
- Set of three 500ml cans
Just add water and a thermometer to each of the three cans, place in the sun or near a bright light bulb and watch the temperatures rise.
Especificaciones
- Description:Radiation Cans, Set of 3