VWR® Buchner Funnels with Fritted Disc
Proveedor: VWR International
Certificados
Acerca de este artículo
Ideal for use where filter paper or membranes cannot withstand chemical attack of the filtrate.
- Borosilicate glass fabrication
- Fritted disc
- Beaded top rims
Funnels feature medium-walled stems and beaded top rims for extra strength.
Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. Funnel meets federal specification DD- F -776.
These funnels are used in qualitative inorganic analysis and preparative chemistry and are resistant to corrosion and heat.
Frequently Bought Together