Antistatic Polystyrene Pour Boats, Simport Scientific
Proveedor: SIMPORT SCIENTIFIC INC.
Certificados
Acerca de este artículo
These three sizes of antistatic polystyrene pour boats are specially made to facilitate dispensing of powdered and liquid materials.
- Smooth, uniform and economical
The molded material used is thicker than conventional weighing dishes. They can be used safely to weight static-affected materials, and will withstand temperatures up to 80°C.
Frequently Bought Together