Orders LinkContactUsLinkComponent
Quantofix Nitrite Test Strips, CTL Scientific Supply Corp
Quantofix Nitrite Test Strips, CTL Scientific Supply Corp
Nitrite 1-80 PPM Test Strips, 0-5-10-25-50-75-100 mg/L
Catalog # 10217-530
Supplier #:  91311
Quantofix Nitrite Test Strips, CTL Scientific Supply Corp
Catalog # 10217-530
CAS Number:  
Quantofix Nitrite Test Strips, CTL Scientific Supply Corp
Quantofix Nitrite Test Strips, CTL Scientific Supply Corp
Nitrite 1-80 PPM Test Strips, 0-5-10-25-50-75-100 mg/L
CAS Number:  
Price
$1,540.56
Log in for availability
Each (1UN)
Product Details & Documents
Quantofix Nitrite test strips for the semi-quantitative determination of nitrite NO2 - in solutions.
Quantofix Nitrite 3000 test strips for the semi-quantitative determination of high concentrations of nitrite (NO 2- ) in solutions. Between pH 2 and 12 the reaction is independent of the pH value of the test solution. Adjust strongly acidic solutions with dilute sodium hydroxide solution and strongly alkaline solutions with dilute sulphuric acid to a pH of about 4 to 6.
Color change white to red-violet for 1-80 ppm strips. Color change yellow to red for 100-3000 ppm strips. Store nitrite test strips in a cool and dry place, protected from sunlight and moisture.
Ordering information: Packaged as a box of 100.
Documents
Specifications
Specifications
  • Description:
    Nitrite 1-80 PPM Test Strips
  • Range:
    0-5-10-25-50-75-100 mg/L
  • Cat. No.:
    10217-530
  • Storage:
    RT