ContactUsLinkComponent
Test Tubes without Rim, Borosilicate Glass, United Scientific Supplies
  470164-056
undefined
Test Tubes without Rim, Borosilicate Glass, United Scientific Supplies
  470164-056
 :  TT9820-G
  • Descripción:
    Test Tubes without Rim, 18 x 150mm, 24 mL, pack of 72
  • Referencia:
    470164-056
  • Nº de proveedor:
    TT9820-G
Pre-cleaned borosilicate glass test tubes without rims.
  • Reusable
  • Autoclaved
Tubes can be used for hot or cold applications.