Flask Stand, PP, United Scientific Supplies
470164-044
- Descripción:Flask Stand, PP
- Referencia:470164-044
- Nº de proveedor:45105
For use with flasks up to 10 L, this polypropylene flask stand is ribbed to safely hold flasks in place.
- Autoclavable
The flask stand has 12 different steps to hold a variety of flasks centerd and stable.