Cobalt Glass Plates, United Scientific Supplies
Improves visibility in sodium and potassium flame tests.
- Uniform in color
- Available in various sizes
High-quality dark blue cobalt glass filter is used in sodium and potassium salt flame tests. When a flame is viewed through this filter, the color orange is blocked so that a violet-colored flame is visible.
Frequently Bought Together