YMC-Pack Pro C18 HPLC Columns, YMC AMERICA, INC
Supplier: YMC AMERICA, INC
Certificates
About this item
These columns use tightly controlled, highly purified silica, a high-coverage C18 bonding, and a unique end-capping procedure utilizing Lewis acid/Lewis base chemistry.
- Made with highly inert, ultrapure, pH neutral silica
- Very high efficiency columns
These features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules, especially basic pharmaceuticals and chelating compounds.
Specifications
- Catalog No:
- 97029-418
- 97029-440
- 97029-406
- 97029-304
- Carga de carbono:
- 16%
- 16%
- 16%
- 16%
- Extremo tapado:
- Yes
- Yes
- Yes
- Yes
- Rango de pH:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8