Lab Spatulas, Stainless Steel, Chemglass
Catalog # 10034-424
Supplier: CHEMGLASS INC.
Small Business Enterprise
Specifications
- Descripción:Spatula, straight scoop, stainless steel
- Longitud:16.5 cm (6.5")
- Referencia:10034-424
- Nº de proveedor:CG-1979-50
Specifications
About this item
Spatula with one pointed end and one blunt end allows for digging out and handling of small amounts of material.