Wall Hooks, White, TrippNT
Triple Hook, Magnet Mount, 33 cm (13"), 5.8 cm (2.25"), 16.5 cm (6.5"), 13.6 kg (30 lbs.)
Catalog # 11376-408
Supplier: TrippNT
Supplier #: 50086
Jump to:
Product Details & Documents
Non-rusting, non-magnetic super strong white lab hooks install instantly with tape or magnet.
- Can hold up to 10 pounds on each hook
- Choose from single, double, triple, quadruple, quintuple
- Great for lab coats and keys
- Easily mounts with tape or magnet
- Unbreakable construction
- American made by TrippNT
- Total replacement Lifetime Guarantee
Sizes range from a single hook up to 5 hooks on one back. Use them individually or install them end-to-end to provide the proper storage. The extra wide tab on the hook offers a wider surface area for items that otherwise might slip. Hooks are excellent for storing lab coats, wire, tubing, in closets, changing areas, offices, entry ways and manufacturing areas.
Documents
Especificaciones
- Description:Triple Hook, Magnet Mount
- Height:16.5 cm (6.5")
- Load Capacity:13.6 kg (30 lbs.)
- Depth:5.8 cm (2.25")
- Width:33 cm (13")