VWR® Unisex 100% Cotton Lab Coat
Supplier: VWR International
Certificates
About this item
These unisex cotton lab coats are made of breathable, comfortable, 100% cotton fabric and feature a quick-release five-snap front and snap cuffs
- Quick-release five-snap front and snap cuffs
- Plain front with no embroidery
- Overall length is 99 cm (39")
In addition, the 100% cotton lab coat features one breast pocket with a pencil slot, plus two oversized hip pockets. Side vents are also included for easy pants pocket access.
Ordering information: Sizes XX-Small, 4X-Large, and 5X-Large are also available via special order. Contact your Avantor sales representative for more information. Please allow up to 6 weeks for delivery of special order items.
Specifications
- Catalog No:
- 10753-936
- 10753-934
- 10753-938
- 10753-932
- Aplicaciones de vestimenta:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Género:
- Para hombres
- Para hombres
- Para hombres
- Para hombres
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Tamaño de cierre:
- Con botones a presión
- Con botones a presión
- Con botones a presión
- Con botones a presión
- Tela:
- Cotton
- Cotton
- Cotton
- Cotton
Specifications
Related Information
Frequently Bought Together