PVC/Polyester Garment, Snap Fly Front, MCR Safety
Catalog # 80082-282
Supplier: MCR SAFETY
Specifications
- Talla de ropa:3XL
- Ajuste de la prenda:Regular
- Tamaño de cierre:Con botones a presión
- Aplicaciones de vestimenta:Industrial,Laboratory
- Género:Unisex
- Color:Yellow
- Tela:PVC/polyester
- Grosor:.35 mm
- Referencia:80082-282
- Nº de proveedor:200BPX3
Specifications
About this item
The Classic Series Material is the most popular in the Industry
It combines the strength and waterproof qualities of PVC with the durability of a polyester backing. The 200BP yellow Bib style pant features partial elastic suspenders, snap fly front, and take up snaps on the ankles and the waist. Come back to the Classics for your protective wear needs.