VWR® Acrylic Dispenser for Parafilm Sealing Film
82024-547
- Descripción:Parafilm Dispenser
- Referencia:82024-547
- An x Prf x Al:119×175×168 mm
Strong, crystal-clear acrylic dispenser makes film supply visible and features a serrated cutting blade.
- Heavy-duty construction for increased durability
- Dispenser holds one or two 50 mm (2") rolls, or one 100 mm (4") roll
Store, dispense, and cut Parafilm, tape, labels, and other compatible rolls.