ContactUsLinkComponent
SHURStain™ Manual Slide Stainers, Triangle Biomedical
SHURStain™ Manual Slide Stainers, Triangle Biomedical
Catalog # 89238-896
undefined
SHURStain™ Manual Slide Stainers, Triangle Biomedical
Catalog # 89238-896
Supplier Number:  SS-6L
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Descripción:
    Manual Stainer Rack, 6 position
  • Referencia:
    89238-896
  • D×W×H:
    160×375×156 mm (6×15×6")
  • Nº de proveedor:
    SS-6L
Specifications
About this item
The SHURStain™ Manual Stainers utilizes customizable manual stainer racks that are designed for the needs of today’s lab - where increased reliability, efficiency, throughput, accuracy and safety are all critical factors for delivering the highest quality care to your patients.
These modular stainers enable you to select the number of individual staining positions necessary to meet your histology, cytology, dermatology, immunohistochemical, hematology, microbiology or special staining application. They are produced in 3, 6 and 12 well configurations, and multiple units can be interlocked backto- back to increase staining volume capabilities. Sliding lids keep stains and reagents covered when not in active use, preventing unwanted evaporation.
The various well configurations allow any style it to easily set on top of croystats to perform staining on frozen sections. Each SHURStain™ rack comes with a supply of replaceable documentation strips to record reagent brand, lot #, date changed, etc. The number of data positions is proportional to the number of wells. Digital Timer comes with bracket attachment, which provides countdown, built-in memory, loud alarm and large display.
includes 2ea (SS-WLW), 1ea (SS-WLG), 1ea (SS-SRG-24), includes 4ea (SS-WLW), 2ea (SS-WLG), 1ea (SS-SRG-24), includes 8ea (SS-WLW), 4ea (SS-WLG), 1ea (SS-SRG-24).